Learn More
BIOHIPPO Mouse PIGR Microsample
SDP
Catalog No. 5000315187
Encompass
Mouse PIGR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIGR in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 g/mL with a sensitivity of 0 048 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PIGR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PIGR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PIGR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15187 Supplier BIOHIPPO Supplier No. BHE15212555
Mouse PIGR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PIGR in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 g/mL with a sensitivity of 0 048 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PIGR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PIGR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PIGR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color