Learn More
Sino Biological Anti-pIgR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50119RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse PIGR (rM PIGR; Catalog#50119-M08H; NP_035212.2; Met 1-Lys 645). PIGR specific IgG was purified by mouse PIGR affinity chromatography.
Catalog No. 50-241-1271