Learn More
American Research Products Inc Mouse Plasma protease C1 inhib
Catalog No. 5000328229
Encompass
Mouse Plasma protease C1 inhibitor (SERPING1) ELISA kit from CUSABIO Detection Range 5 6 ng/mL-360 ng/mL Sensitivity 1 4 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28229 Supplier American Research Products Inc Supplier No. CSBEL021086MO
Add to Cart
Add to Cart
Mouse Plasma protease C1 inhibitor (SERPING1) ELISA kit from CUSABIO Detection Range 5 6 ng/mL-360 ng/mL Sensitivity 1 4 ng/mL Sample serum plasma tissue homogenates Method Sandwich