Learn More
American Research Products Inc Mouse plasminogen actiVator in
Catalog No. 5000326504
Encompass
Mouse plasminogen actiVator inhibitor 1 PAI-1 ELISA Kit from CUSABIO Detection Range 0 9 ng/mL-60 ng/mL Sensitivity 0 22 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26504 Supplier American Research Products Inc Supplier No. CSBE07947M
Mouse plasminogen actiVator inhibitor 1 PAI-1 ELISA Kit from CUSABIO Detection Range 0 9 ng/mL-60 ng/mL Sensitivity 0 22 ng/mL Sample serum plasma tissue homogenates Method Sandwich