Learn More
American Research Products Inc Mouse platelet actiVating fact
Catalog No. 5000328936
Encompass
Mouse platelet actiVating factor PAF ELISA KIT from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28936 Supplier American Research Products Inc Supplier No. CSBE08199M
Mouse platelet actiVating factor PAF ELISA KIT from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich