Learn More
Biomatik Corporation Mouse Platelet Derived Growth Factor Receptor Like Protein (PDGFRL) CLIA Kit
Supplier:  Biomatik Corporation EKU09547
Encompass_Preferred
Mouse Platelet Derived Growth Factor Receptor Like Protein (PDGFRL) CLIA Kit for detecting PDGFRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.86-5,000pg/ml. Sensitivity: 2.47pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q6PE55. Gene ID: 68797. Target Synonyms: PRLTS; PDGF-Receptor Beta-Like Tumor Suppressor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5899