Learn More
American Research Products Inc Mouse platelet deriVed growth
Catalog No. 5000329510
Encompass
Mouse platelet-deriVed growth factor-BB PDGF-BB ELISA Kit from CUSABIO Detection Range 0 18 ng/mL-12 ng/mL Sensitivity 0 047 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29510 Supplier American Research Products Inc Supplier No. CSBE08925M
Mouse platelet-deriVed growth factor-BB PDGF-BB ELISA Kit from CUSABIO Detection Range 0 18 ng/mL-12 ng/mL Sensitivity 0 047 ng/mL Sample serum plasma tissue homogenates Method Sandwich