Learn More
American Research Products Inc Mouse platelet deriVed growth
Catalog No. 5000328408
Encompass
Mouse platelet-deriVed growth factor CC (PDGF-CC) ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28408 Supplier American Research Products Inc Supplier No. CSBE13404M
Mouse platelet-deriVed growth factor CC (PDGF-CC) ELISA Kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich