Learn More
American Research Products Inc Mouse Platelet deriVed growth
Catalog No. 5000327997
Encompass
Mouse Platelet-deriVed growth factor subunit B (PDGFB) ELISA kit from CUSABIO Detection Range 93 75 pg/mL-6000 pg/mL Sensitivity 23 44 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27997 Supplier American Research Products Inc Supplier No. CSBEL017709MO
Mouse Platelet-deriVed growth factor subunit B (PDGFB) ELISA kit from CUSABIO Detection Range 93 75 pg/mL-6000 pg/mL Sensitivity 23 44 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich