Learn More
American Research Products Inc Mouse Platelet deriVed growth
Catalog No. 5000328026
Encompass
Mouse Platelet-deriVed growth factor DD (PDGF-DD) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28026 Supplier American Research Products Inc Supplier No. CSBEL017711MO
Mouse Platelet-deriVed growth factor DD (PDGF-DD) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich