Learn More
Biomatik Corporation Mouse Platelet Derived Growth Factor Receptor Like Protein (PDGFRL) ELISA Kit
Supplier:  Biomatik Corporation EKU06695
Encompass_Preferred
Mouse Platelet Derived Growth Factor Receptor Like Protein (PDGFRL) ELISA Kit for detecting PDGFRL in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.12-5000pg/ml. Sensitivity: 30.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6PE55. Gene ID: 68797. Target Synonyms: PRLTS; PDGF-Receptor Beta-Like Tumor Suppressor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6039