Learn More
Biomatik Corporation Mouse Platelet Derived Growth Factor BB (PDGF BB) ELISA Kit
Supplier:  Biomatik Corporation EKU11917
Encompass_Preferred
Mouse Platelet Derived Growth Factor BB (PDGF BB) ELISA Kit for detecting PDGF BB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.071ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31240. Gene ID: 18591. Target Synonyms: PDGF-BB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6668