Learn More
American Research Products Inc Mouse Platelet DeriVed Growth
Catalog No. 5000329141
Encompass
Mouse Platelet-DeriVed Growth Factor Soluble Receptor a PDGFsR-a ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29141 Supplier American Research Products Inc Supplier No. CSBE04699M
Mouse Platelet-DeriVed Growth Factor Soluble Receptor a PDGFsR-a ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich