Learn More
American Research Products Inc Mouse Platelet Factor 4 PF 4
Catalog No. 5000326529
Encompass
Mouse Platelet Factor 4 PF-4 ELISA Kit from CUSABIO Detection Range 3 125 ng/mL-200 ng/mL Sensitivity 1 603 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26529 Supplier American Research Products Inc Supplier No. CSBE07884M
Mouse Platelet Factor 4 PF-4 ELISA Kit from CUSABIO Detection Range 3 125 ng/mL-200 ng/mL Sensitivity 1 603 ng/mL Sample serum plasma tissue homogenates Method Sandwich