Learn More
American Research Products Inc Mouse Platelet glycoprotein 4
Catalog No. 5000327430
Encompass
Mouse Platelet glycoprotein 4 (CD36) ELISA kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9pg/ml Sample serum plasma tissue homogenates Method
Catalog No. 50-003-27430 Supplier American Research Products Inc Supplier No. CSBEL004927MO
Mouse Platelet glycoprotein 4 (CD36) ELISA kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9pg/ml Sample serum plasma tissue homogenates Method