Learn More
Biomatik Corporation Mouse Platelet/Endothelial Cell Adhesion Molecule (PECAM1) ELISA Kit
Supplier:  Biomatik Corporation EKU06711
Encompass_Preferred
Mouse Platelet/Endothelial Cell Adhesion Molecule (PECAM1) ELISA Kit for detecting PECAM1 in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q08481. Gene ID: 18613. Target Synonyms: CD31; EndoCAM; GPIIA'; PECA1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7189