Learn More
Sino Biological Recombinant Anti-uPAR/PLAUR Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50160R014100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse PLAUR / CD87 (rM PLAUR / CD87; Catalog#50160-M08H; NP_035243.1; Met1-Thr297).
Catalog No. 50-240-8952