Learn More
Sino Biological Anti-uPAR/PLAUR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50160RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse PLAUR extracellular domain (rM PLAUR; Catalog#50160-M08H; NP_035243.1; Met 1-Thr 297). PLAUR specific IgG was purified by mouse PLAUR affinity chromatography.
Catalog No. 50-241-2610