Learn More
Raybiotech Inc Mouse PLGF-2 ELISA Kit
Supplier:  Raybiotech Inc ELMPLGF21
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 1 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The sandwich ELISA antibody pair detects Mouse PLGF-2.
Catalog No. 50-195-0114