Learn More
BIOHIPPO Mouse PLTP Microsample
SDP
Catalog No. 5000315833
Encompass
Mouse PLTP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PLTP in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PLTP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PLTP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PLTP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15833 Supplier BIOHIPPO Supplier No. BHE15210858
Mouse PLTP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PLTP in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 062 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PLTP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PLTP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PLTP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color