Learn More
BIOHIPPO Mouse PMP22 Microsample
SDP
Catalog No. 5000313951
Encompass
Mouse PMP22 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PMP22 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 059 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PMP22 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PMP22 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PMP22 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13951 Supplier BIOHIPPO Supplier No. BHE15211088
Mouse PMP22 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PMP22 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 059 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PMP22 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PMP22 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PMP22 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color