Learn More
Biomatik Corporation Mouse Polyamine Oxidase (PAOX) ELISA Kit
Supplier:  Biomatik Corporation EKU06737
Encompass_Preferred
Mouse Polyamine Oxidase (PAOX) ELISA Kit for detecting PAOX in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.12-5000pg/ml. Sensitivity: 32pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8C0L6. Gene ID: 212503. Target Synonyms: PAO; Peroxisomal N (1)-Acetyl-Spermine/Spermidine Oxidase (Exo-N4-Amino). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4713