Learn More
Abcam Mouse on Mouse Polymer IHC Kit, 15ML
Catalog No. NC1937491
Encompass_Preferred
Mouse on mouse IHC kit allows antibodies raised in mouse to be used on mouse tissues in IHC, without high backgrounds caused by anti-mouse secondary antibody binding to endogenous mouse antibodies in mouse tissues. - Cited in over 10 publications - HRP polymer for strong, specific detection - Minimal cross-reactivity with human/rabbit - Compatible with standard DAB staining
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1937491 Supplier Abcam Supplier No. AB26945215ML
Mouse on mouse IHC kit allows antibodies raised in mouse to be used on mouse tissues in IHC, without high backgrounds caused by anti-mouse secondary antibody binding to endogenous mouse antibodies in mouse tissues. - Cited in over 10 publications - HRP polymer for strong, specific detection - Minimal cross-reactivity with human/rabbit - Compatible with standard DAB staining
The product is subject to the following: Abcam Restricted Use Statement