Learn More
Active Motif Mouse Positive Control Primer Set Gapdh-2
SDP
Catalog No. NC0235917
Encompass
Mouse Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with mouse samples. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP samples from multiple mouse cell lines.
Catalog No. NC0235917 Supplier Active Motif Supplier No. 71018
Mouse Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with mouse samples. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP samples from multiple mouse cell lines.