Learn More
Active Motif Mouse Positive Control Primer Set Grik3
SDP
Catalog No. NC1850755
Encompass
Mouse Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with mouse samples. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP samples from multiple mouse cell lines.
Catalog No. NC1850755 Supplier Active Motif Supplier No. 71021
Mouse Control qPCR Primer Sets were designed to serve as positive or negative ChIP controls when performing chromatin immunoprecipitation with mouse samples. Each primer set is a mixture of forward and reverse primers that have been validated for qPCR and endpoint PCR of ChIP samples from multiple mouse cell lines.