Learn More
Sino Biological Anti-Periostin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50450RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Periostin / POSTN (rM Periostin / POSTN; Catalog#50450-M08H; NP_056599.1; Met 1-Gln 811). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-9498