Learn More
Sino Biological Mouse PPARA Protein 50ug (Sino Biological)
Supplier:  Sino Biological 56956M07E50
Encompass_Preferred
A DNA sequence encoding the Mouse PPARA (P23204) (Thr200-Tyr468) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-2956