Learn More
BIOHIPPO Mouse PPARd Microsample
SDP
Catalog No. 5000314861
Encompass
Mouse PPARd Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PPARd in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 057 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PPARd Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PPARd Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PPARd biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14861 Supplier BIOHIPPO Supplier No. BHE15210957
Mouse PPARd Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PPARd in tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 057 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PPARd Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PPARd Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PPARd biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color