Learn More
Sino Biological Anti-PPAR gamma /NR1C3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 107445T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Mouse PPAR gamma /NR1C3, and purified by antigen affinity chromatography.
Catalog No. 50-255-5182