Learn More
Sino Biological Anti-CXCL7/NAP-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50145T32100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CXCL7/PPBP (rM CXCL7/PPBP; Catalog#50145-M07E; Q9EQI5; Ile49-Ile109). CXCL7/PPBP specific IgG was purified by Mouse CXCL7/PPBP affinity chromatography.
Catalog No. 50-255-3413