Learn More
Sino Biological Anti-PPM1A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50277RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse PPM1A (rM PPM1A; Catalog#50277-M08E; NP_032936.1; Met 1-Trp 382). PPM1A specific IgG was purified by mouse PPM1A affinity chromatography.
Catalog No. 50-241-1286