Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse PRDC (GREM2) ELISA Kit

Catalog No. EMGREM2X5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EMGREM2X5 5 x 96 Tests
EMGREM2 96 Tests
EMGREM2X10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EMGREM2X5 Supplier Invitrogen™ Supplier No. EMGREM2X5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Mouse PRDC (GREM2) ELISA quantitates Ms PDRC in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms PDRC. Principle of the method The Mouse PDRC solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

GREM2 encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted glycosylated protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O88273
Assay Range 3.28-800 pg/mL
Assay Sensitivity 3 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Other Proteins
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias CKTSF1B2; CKTSF1B2FLJ21195; Cysteine knot superfamily 1, BMP antagonist 2; DAN domain family member 3; DAND3; DAND3Prdc; GREM2; gremlin 2; gremlin 2 homolog, cysteine knot superfamily; gremlin 2 homolog, cysteine knot superfamily (Xenopus laevis); gremlin 2, cysteine knot superfamily, homolog; gremlin 2, DAN family BMP antagonist; Gremlin1; Gremlin2; gremlin-2; Prdc; PRDCgremlin 2; Protein related to DAN and cerberus; RGD1560008CKTSF1B2; CKTSF1B2FLJ21195; Cysteine knot superfamily 1, BMP antagonist 2; DAN domain family member 3; DAND3; DAND3Prdc; GREM2; gremlin 2; gremlin 2 homolog, cysteine knot superfamily; gremlin 2 homolog, cysteine knot superfamily (Xenopus laevis); gremlin 2, cysteine knot superfamily, homolog; gremlin 2, DAN family BMP antagonist; Gremlin1; Gremlin2; gremlin-2; Prdc; PRDCgremlin 2; Protein related to DAN and cerberus; RGD1560008; Gremlin2; Prdc
Gene ID (Entrez) 23893
Gene Symbol Grem2
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Cysteine knot superfamily 1, BMP antagonist 2, Protein related to DAN and cerberus, PRDC
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 33 μL, Serum 33 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target GREM2
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.