Learn More
ACROBiosystems Monoclonal Anti-pre-Gc protein (SFTS virus (HN6)) Antibody, Mouse IgG1 (13E1)
Catalog No. 502719046
Encompass_Preferred
Immobilized SFTS virus pre-Gc Protein (HN6), His Tag (Cat. No. PRN-S52H3) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-pre-Gc protein (SFTS virus (HN6)) Antibody, Mouse IgG1 (13E1) (Cat. No. PRN-Y315) with a linear range of 0.1-2 ng/mL (QC tested).
Catalog No. 50-271-9046 Supplier ACROBiosystems Supplier No. PRNY3151MG
Immobilized SFTS virus pre-Gc Protein (HN6), His Tag (Cat. No. PRN-S52H3) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-pre-Gc protein (SFTS virus (HN6)) Antibody, Mouse IgG1 (13E1) (Cat. No. PRN-Y315) with a linear range of 0.1-2 ng/mL (QC tested).