Learn More
Abcam Mouse Prealbumin ELISA Kit, 1 x 384 Tests
Catalog No. 502655395
Encompass_Preferred
Mouse Prealbumin ELISA Kit is a single-wash 90-min SimpleStep Sandwich ELISA used to quantify Mouse Prealbumin (Transthyretin, TTR) with a sensitivity of 62.48 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout, works on any plate reader - Different formats for different needs: 384-well for higher throughput - Need to design your own assay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-5395 Supplier Abcam Supplier No. AB2822971X384TESTS
Add to Cart
Add to Cart
Mouse Prealbumin ELISA Kit is a single-wash 90-min SimpleStep Sandwich ELISA used to quantify Mouse Prealbumin (Transthyretin, TTR) with a sensitivity of 62.48 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout, works on any plate reader - Different formats for different needs: 384-well for higher throughput - Need to design your own assay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement