Learn More
Raybiotech Inc Mouse Pref-1/DLK-1/FA1 ELISA Kit
Supplier:  Raybiotech Inc ELMPREF11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.5 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Mouse Pref-1.
Catalog No. 50-195-0116