Learn More
Cayman Chemical Mouse Pregnane X Receptor Reporter Assay System, 96 Well
SDP
Catalog No. 502937386
Encompass_Preferred
mPXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.[INDIGO Catalog Nos. M07001-32, M07001]
Catalog No. 50-293-7386 Supplier Cayman Chemical Supplier No. 2834296 WELL
mPXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.[INDIGO Catalog Nos. M07001-32, M07001]