Learn More
Biomatik Corporation Mouse Prekallikrein (PK) ELISA Kit
Supplier:  Biomatik Corporation EKU11971
Encompass_Preferred
Mouse Prekallikrein (PK) ELISA Kit for detecting PK in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P26262. Gene ID: 16621. Target Synonyms: KLKB1; KLK3; Fletcher Factor; Kallikrein B,Plasma; Kininogenin; Plasma prekallikrein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8058