Learn More
Biomatik Corporation Mouse Prenylcysteine Oxidase 1 (PCYOX1) ELISA Kit
Supplier:  Biomatik Corporation EKU06759
Encompass_Preferred
Mouse Prenylcysteine Oxidase 1 (PCYOX1) ELISA Kit for detecting PCYOX1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.072ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9CQF9. Gene ID: 66881. Target Synonyms: PCL1; Prenylcysteine lyase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8809