Learn More
BIOHIPPO Mouse Presepsin Microsample
SDP
Catalog No. 5000315096
Encompass
Mouse Presepsin Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Presepsin in Serum plasma and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 4 72 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse Presepsin Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse Presepsin Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse Presepsin biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15096 Supplier BIOHIPPO Supplier No. BHE15210595
Mouse Presepsin Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Presepsin in Serum plasma and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 4 72 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse Presepsin Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse Presepsin Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse Presepsin biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color