Learn More
BIOHIPPO Mouse PRG4 Microsample
SDP
Catalog No. 5000314656
Encompass
Mouse PRG4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PRG4 in serum plasma synovial fluid and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 066 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PRG4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PRG4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PRG4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14656 Supplier BIOHIPPO Supplier No. BHE15210779
Mouse PRG4 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PRG4 in serum plasma synovial fluid and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 066 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse PRG4 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse PRG4 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse PRG4 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color