Learn More
Sino Biological Anti-Prolactin Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50457T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Prolactin Receptor (rM Prolactin Receptor; Catalog#50457-M08H; P07321; Met1-Asp229). Prolactin Receptor specific IgG was purified by Mouse Prolactin Receptor affinity chromatography.
Catalog No. 50-255-3271