Learn More
American Research Products Inc Mouse pro interleukin 1 beta
Catalog No. 5000326274
Encompass
Mouse pro interleukin 1 beta (pro-IL1B) ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26274 Supplier American Research Products Inc Supplier No. CSBET011614MO
Mouse pro interleukin 1 beta (pro-IL1B) ELISA Kit from CUSABIO Detection Range 6 25 pg/mL-400 pg/mL Sensitivity 1 56 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich