Learn More
BIOHIPPO Mouse Pro-MMP-9 Microsample
SDP
Catalog No. 5000314844
Encompass
Mouse Pro-MMP-9 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Pro-MMP-9 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 478 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse Pro-MMP-9 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse Pro-MMP-9 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse Pro-MMP-9 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14844 Supplier BIOHIPPO Supplier No. BHE15210690
Mouse Pro-MMP-9 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of Pro-MMP-9 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 79-50 ng/mL with a sensitivity of 0 478 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse Pro-MMP-9 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse Pro-MMP-9 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse Pro-MMP-9 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color