Learn More
American Research Products Inc Mouse Pro NerVe growth factor
Catalog No. 5000327887
Encompass
Mouse Pro-NerVe growth factor (proNGF) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27887 Supplier American Research Products Inc Supplier No. CSBEQ027721MO
Mouse Pro-NerVe growth factor (proNGF) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich