Learn More
American Research Products Inc Mouse Pro opiomelanocortin PO
Catalog No. 5000327231
Encompass
Mouse Pro-opiomelanocortin (POMC) ELISA KIT from CUSABIO Detection Range 46 9 pg/ml-3000 pg/ml Sensitivity 24 pg/ml Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-27231 Supplier American Research Products Inc Supplier No. CSBEL018363MO
Mouse Pro-opiomelanocortin (POMC) ELISA KIT from CUSABIO Detection Range 46 9 pg/ml-3000 pg/ml Sensitivity 24 pg/ml Sample serum plasma cell culture supernates tissue homogenates Method Competitive