Learn More
Abcam Mouse Pro-Collagen I alpha 1 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2105791X96TESTS
Encompass_Preferred
Mouse Pro-Collagen I alpha 1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse Pro-Collagen I alpha 1 with a sensitivity of 6.7 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair - Cited in over 10 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1882100