Learn More
Biomatik Corporation Mouse Procollagen III (PCIII) ELISA Kit
Supplier:  Biomatik Corporation EKU11361
Encompass_Preferred
Mouse Procollagen III (PCIII) ELISA Kit for detecting PCIII in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.65ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: PCOL-III; PC-III; PC3; Type III Procollagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7057