Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse Progesterone Rapid ELISA Kit

Catalog No. EELR011
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EELR011 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EELR011 Supplier Invitrogen™ Supplier No. EELR011
Add to Cart
Add to Cart

ELISA

The Mouse Progesterone (Pg) ELISA quantitates Pg in serum and plasma. Principle of the method The Mouse Pg solid phase competitive ELISA (enzyme linked immunosorbent assay) is designed to measure the amount of the target bound between the pre coated antigen and the HRP linked antibody specific for Mouse Pg. A target specific antigen has been pre coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells, where Mouse Pg in the samples (or standards) competes with the immobilized (capture) antigen for sites on the HRP linked detection antibody. Excess conjugate and unbound sample or standard are washed away from the plate. The substrate solution is added to each well, and the enzyme substrate reaction is terminated by the addition of a stop solution, turning the color yellow. The optical density (OD) is measured spectrophotometrically at a wavelength of 450±2 nm. The intensity of this signal is inversely proportional to the concentration of Mouse Pg present in the original specimen, which is determined by comparing the OD of the samples to the standard curve. Achieve fast and reliable results in just 90 minutes with our Rapid ELISA kit, featuring a streamlined protocol that requires only one wash step. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation...

Progesterone is the major progestational steroid, secreted primarily by the corpus luteum and the placenta. Progesterone binds to the progesterone receptor, resulting in dissociation of heat shock proteins, receptor phosphorylation, and transcription activation through direct or indirect interaction with transcription factors. Progesterone acts on the uterus, the mammary glands and the brain. It is required in embryo implantation, pregnancy maintenance, and the development of mammary tissue for milk production. Progesterone, converted from pregnenolone, also serves as an intermediate in the biosynthesis of gonadal steroid hormones and adrenal corticosteroids. Therapeutic progesterone is a synthetic form of the endogenous hormone progesterone. This agent exerts inhibitory effects on estrogens by decreasing the number of estrogen receptors and increasing its metabolism to inactive metabolites. [National Center for Biotechnology Information. PubChem Compound Database; CID=5994].
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 78.13-5000 pg/mL
Assay Sensitivity 24.97 pg/mL
Conjugate HRP
Immunoassay Kit Format Coated ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum
Target Species Mouse
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 11-alpha-hydroxyprogesterone; Agolutin; C21H30O2; Crinone; Luteohormone; OHP; Pregn-4-ene-3; Progesterone-11a-hemisuccinate11-alpha-hydroxyprogesterone; Agolutin; C21H30O2; Crinone; Luteohormone; OHP; Pregn-4-ene-3; Progesterone-11a-hemisuccinate;
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, HRP Conjugate, Standard & Sample Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, HRP Conjugate, Standard & Sample Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Progesterone
Test Time 15 min
Total Assay Time 1 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.