Learn More
American Research Products Inc Mouse Prokineticin 2 PROK2 E
Catalog No. 5000327157
Encompass
Mouse Prokineticin-2 (PROK2) ELISA kit from CUSABIO Detection Range 0 68 ng/ml - 500 ng/ml Sensitivity 0 68 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27157 Supplier American Research Products Inc Supplier No. CSBEL018747MO
Mouse Prokineticin-2 (PROK2) ELISA kit from CUSABIO Detection Range 0 68 ng/ml - 500 ng/ml Sensitivity 0 68 ng/ml Sample serum plasma tissue homogenates Method Sandwich