Learn More
Sino Biological Mouse PROS1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 68097MM18320
Encompass_Preferred
A DNA sequence encoding the Mouse PROS1 (Q08761) (Met1-Phe675) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-4534